These REDSTICK Box Levels offer industry-leading performance with best-in-class vial readability and frame durability. Featuring a REINFORCED BACKBONE for a strong, durable frame and SHARPSITE Vial Technology for optimal readability with a magnified bubble and high-visibility spirit.
- Reinforced backbone for strongest, most durable frame with reinforced vial retention.
- Durable, all-metal hang hole for storage.
- Guaranteed lifetime accuracy: .029° (.0005"/in, 0.5mm/M).
- High contrast vial system for easy cleaning and optimized visibility.
- High density, removable end-caps protect frame and provide access in tight spaces.
- Overmolded, non-slip grips prevent sliding.
- Precision milled measuring surface ensures accuracy in all working positions.
- Sharpsite vial technology offers best readability and impact protection.
- Wide, durable handles for comfort and portability.