Learn More
MSC Milwaukee Tool Class 2 Breakaway Safety Vest, 4XL/5XL, 1 ea
Catalog No. 5000770164
Encompass
This ANSI Type R Class 2 high visibility yellow mesh safety vest is designed for enhanced visibility and comfort in demanding work environments. It features moisture-wicking and anti-microbial treated fabric to keep the wearer cooler and reduce odor, along with a lightweight, breathable mesh construction.
  • Lightweight, breathable mesh fabric
  • External size adjustment for a better fit
  • Five-point breakaway design on shoulders, sides, and front for safety
  • Clear ID holder with hook-and-loop closure
  • Pass-through slit for fall protection gear
  • Reinforced pen pockets made with 600D tear-resistant material
  • Durable dual mic tabs
  • One internal pocket for storage
  • Hang loop for easy storage
  • Third-party tested for reliability
  • ANSI 107-2020 Class 2 Type R compliant
Catalog No. 50-007-70164 Supplier MSC Supplier No. 10882702
This ANSI Type R Class 2 high visibility yellow mesh safety vest is designed for enhanced visibility and comfort in demanding work environments. It features moisture-wicking and anti-microbial treated fabric to keep the wearer cooler and reduce odor, along with a lightweight, breathable mesh construction.
  • Lightweight, breathable mesh fabric
  • External size adjustment for a better fit
  • Five-point breakaway design on shoulders, sides, and front for safety
  • Clear ID holder with hook-and-loop closure
  • Pass-through slit for fall protection gear
  • Reinforced pen pockets made with 600D tear-resistant material
  • Durable dual mic tabs
  • One internal pocket for storage
  • Hang loop for easy storage
  • Third-party tested for reliability
  • ANSI 107-2020 Class 2 Type R compliant