Learn More
MSC Milwaukee Tool Cut-Resistant Gloves, ANSI A2, Medium, 1 Pair
Catalog No. 5000796375
Encompass
These high-visibility, cut-resistant gloves offer enhanced protection and dexterity for various tasks. Featuring a reinforced nitrile coating on the palm and fingers, they provide durability in high-wear areas and include Smartswipe™ touchscreen-compatible fingertips. The 15-gauge knit ensures a comfortable and breathable fit, promoting better dexterity for handling small objects.
  • Designed for enhanced fit and protection
  • ANSI/ISEA A2 cut resistance level
  • Reinforced nitrile coating for durability
  • Features a 15 gauge knit for superior dexterity
  • Lightweight and breathable for all-day comfort
  • Smartswipe™ touchscreen compatible fingertips
  • High performance polyethylene provides cut protection
  • EN388 rated: abrasion 3, cut 2, tear 4, puncture 2, TDM cut B
Catalog No. 50-007-96375 Supplier MSC Supplier No. 15095433
These high-visibility, cut-resistant gloves offer enhanced protection and dexterity for various tasks. Featuring a reinforced nitrile coating on the palm and fingers, they provide durability in high-wear areas and include Smartswipe™ touchscreen-compatible fingertips. The 15-gauge knit ensures a comfortable and breathable fit, promoting better dexterity for handling small objects.
  • Designed for enhanced fit and protection
  • ANSI/ISEA A2 cut resistance level
  • Reinforced nitrile coating for durability
  • Features a 15 gauge knit for superior dexterity
  • Lightweight and breathable for all-day comfort
  • Smartswipe™ touchscreen compatible fingertips
  • High performance polyethylene provides cut protection
  • EN388 rated: abrasion 3, cut 2, tear 4, puncture 2, TDM cut B