Learn More
MSC Milwaukee Tool Cut-Resistant Gloves, ANSI/ISEA A2, Small, 1 Pair
Catalog No. 5000674951
Encompass
These cut-resistant gloves are designed for demanding applications, featuring a durable polyethylene and nitrile-coated construction for enhanced protection and grip.
  • ANSI/ISEA A2 cut resistance
  • Nitrile-coated palm and fingers
  • Nitrile dipped grip surface
  • Fine 18 gauge knit for dexterity
  • Elastic cuff style for secure fit
  • Small size (numeric 7)
  • Overall length of 10 in
Catalog No. 50-006-74951 Supplier MSC Supplier No. 51347060
These cut-resistant gloves are designed for demanding applications, featuring a durable polyethylene and nitrile-coated construction for enhanced protection and grip.
  • ANSI/ISEA A2 cut resistance
  • Nitrile-coated palm and fingers
  • Nitrile dipped grip surface
  • Fine 18 gauge knit for dexterity
  • Elastic cuff style for secure fit
  • Small size (numeric 7)
  • Overall length of 10 in