Learn More
MSC Milwaukee Tool Cut-Resistant Gloves, Cut A5, Size Small, 1 Pair
Catalog No. 5000677851
Encompass
These cut-resistant gloves provide superior protection for various tasks. They are designed with a high cut resistance level and feature a durable coating for enhanced grip and dexterity.
  • ANSI/ISEA Cut Resistance Level A5
  • ANSI/ISEA Puncture Resistance Level 0
  • ANSI/ISEA Abrasion Resistance Level 0
  • Thirteen gauge construction for better dexterity
  • Small size (Numeric Size 7)
  • Overall length of 9-1/2 in.
  • Polyester and HPPE blend material with polyethylene coating
  • Palm and fingers coating provides a rough grip
  • Touchscreen compatible fingertips
  • Seamless lining
  • Unisex design
Catalog No. 50-006-77851 Supplier MSC Supplier No. 10884005
These cut-resistant gloves provide superior protection for various tasks. They are designed with a high cut resistance level and feature a durable coating for enhanced grip and dexterity.
  • ANSI/ISEA Cut Resistance Level A5
  • ANSI/ISEA Puncture Resistance Level 0
  • ANSI/ISEA Abrasion Resistance Level 0
  • Thirteen gauge construction for better dexterity
  • Small size (Numeric Size 7)
  • Overall length of 9-1/2 in.
  • Polyester and HPPE blend material with polyethylene coating
  • Palm and fingers coating provides a rough grip
  • Touchscreen compatible fingertips
  • Seamless lining
  • Unisex design