These cut-resistant gloves are designed for applications requiring high dexterity and superior cut protection. They feature an 18-gauge construction, providing total mobility for intricate tasks.
- Offers total mobility with 18-gauge construction
- Reinforced nitrile coating between thumb and index finger for added durability
- Long-lasting nitrile dip ensures extended glove life and consistent grip
- Smartswipe™ touchscreen compatible fingertips for device interaction
- High dexterity design ideal for handling small objects
- Breathable glove design enhances comfort during prolonged use
- High performance polyethylene and tungsten knit into gloves for robust cut protection
- Color-coded band for easy cut level identification
- Rough grip surface provides secure handling of materials
- Seamless lining contributes to comfort and reduces irritation