Learn More
MSC Milwaukee Tool Earmuffs, Hard Hat, 26 dB NRR, 1 ea
Catalog No. 5000676713
Encompass
These hard hat mounted earmuffs provide 26 dB noise reduction, designed for compatibility with safety helmets and hard hats.
  • Hard hat mounted type for secure attachment
  • Adjustable fit with center-mount design
  • Comfortable, pressure reducing cushions
  • Can be worn with additional accessories simultaneously
  • Secures into safety helmets and hard hats with BOLT™ and universal accessory slots
Catalog No. 50-006-76713 Supplier MSC Supplier No. 97391627
These hard hat mounted earmuffs provide 26 dB noise reduction, designed for compatibility with safety helmets and hard hats.
  • Hard hat mounted type for secure attachment
  • Adjustable fit with center-mount design
  • Comfortable, pressure reducing cushions
  • Can be worn with additional accessories simultaneously
  • Secures into safety helmets and hard hats with BOLT™ and universal accessory slots