Learn More
MSC Milwaukee Tool Face Shield, Gray, ANSI Z87.1 Rated, 1 ea
Catalog No. 5000767921
Encompass
The BOLT™ Full Face Shield provides extended visibility and robust protection for professional use. Constructed from high-impact polycarbonate, it features both fog-free interior and anti-scratch exterior coatings. This shield integrates seamlessly with the BOLT™ accessory system, allowing quick attachment to compatible head protection.
  • Long-lasting visibility with dual coatings
  • High-impact polycarbonate construction
  • Quick attachment via BOLT™ system
  • Compatible with safety helmets and hard hats
  • ANSI/ISEA Z87.1 (+) - 2020 & CSA Z94.3 rated
  • Offers 180° field of view and UV protection
Catalog No. 50-007-67921 Supplier MSC Supplier No. 78799913
The BOLT™ Full Face Shield provides extended visibility and robust protection for professional use. Constructed from high-impact polycarbonate, it features both fog-free interior and anti-scratch exterior coatings. This shield integrates seamlessly with the BOLT™ accessory system, allowing quick attachment to compatible head protection.
  • Long-lasting visibility with dual coatings
  • High-impact polycarbonate construction
  • Quick attachment via BOLT™ system
  • Compatible with safety helmets and hard hats
  • ANSI/ISEA Z87.1 (+) - 2020 & CSA Z94.3 rated
  • Offers 180° field of view and UV protection