Learn More
MSC Milwaukee Tool Face Shield Lenses, Gray, ANSI Z87.1, 10 pk
Catalog No. 5000789401
Encompass
Full face shields are engineered for long-lasting visibility, featuring ANSI/ISEA Z87.1 (+) - 2020 compliance. Each shield includes a fog-free interior coating and an anti-scratch exterior hard coating to ensure extended product life. Attachment and removal are tool-free with secure snap-in snap-out covers.
  • Long-lasting visibility
  • Fog-free interior coating
  • Anti-scratch exterior hard coating
  • Quick attachment and removal
  • Compatible with safety helmets with no brim
  • High-impact polycarbonate shield
  • Wide 180° field of view
  • Compatible with safety glasses
  • Secure locking detents
Catalog No. 50-007-89401 Supplier MSC Supplier No. 78799921
Full face shields are engineered for long-lasting visibility, featuring ANSI/ISEA Z87.1 (+) - 2020 compliance. Each shield includes a fog-free interior coating and an anti-scratch exterior hard coating to ensure extended product life. Attachment and removal are tool-free with secure snap-in snap-out covers.
  • Long-lasting visibility
  • Fog-free interior coating
  • Anti-scratch exterior hard coating
  • Quick attachment and removal
  • Compatible with safety helmets with no brim
  • High-impact polycarbonate shield
  • Wide 180° field of view
  • Compatible with safety glasses
  • Secure locking detents