Learn More
MSC Milwaukee Tool Face Shield Lenses, Gray Polycarbonate, 10 Pack
Catalog No. 5000766600
Encompass
Full face shields provide long-lasting visibility. These ANSI/ISEA Z87.1 (+) - 2020 rated shields feature a fog-free interior coating and an anti-scratch exterior hard coating for extended product lifetimes. Attachment and removal are simple and tool-free with secure snap-in snap-out covers. Compatible with the BOLT™ accessory system, allowing simultaneous accessory securing. Mount is sold separately.
  • Long-lasting visibility with fog-free interior and anti-scratch exterior
  • Quick attachment and removal; BOLT™ system compatible
  • Includes 10 gray dual coat replacement shields
  • ANSI/ISEA Z87.1 (+) - 2020 & CSA Z94.3 rated
  • High-impact polycarbonate shield with 180° field of view
  • Reusable and provides UV protection
Catalog No. 50-007-66600 Supplier MSC Supplier No. 79944328
Full face shields provide long-lasting visibility. These ANSI/ISEA Z87.1 (+) - 2020 rated shields feature a fog-free interior coating and an anti-scratch exterior hard coating for extended product lifetimes. Attachment and removal are simple and tool-free with secure snap-in snap-out covers. Compatible with the BOLT™ accessory system, allowing simultaneous accessory securing. Mount is sold separately.
  • Long-lasting visibility with fog-free interior and anti-scratch exterior
  • Quick attachment and removal; BOLT™ system compatible
  • Includes 10 gray dual coat replacement shields
  • ANSI/ISEA Z87.1 (+) - 2020 & CSA Z94.3 rated
  • High-impact polycarbonate shield with 180° field of view
  • Reusable and provides UV protection