Learn More
MSC Milwaukee Tool Full Face Shield Lens, Clear Uncoated, 1 ea
Catalog No. 5000768096
Encompass
The Milwaukee Tool Uncoated Full Face Shield Replacement Lens is designed for use with the BOLT™ Task Full Face Shield Frame, providing protection exceeding ANSI/ISEA Z87.1 (+) - 2020 and CSA Z94.3 standards when used together.
  • Contoured fit
  • For use with BOLT™ Task Full Face Shield Frame
  • Includes 1 clear uncoated replacement shield
  • Offers 180° field of view
  • Compatible with safety glasses
  • Tool free shield change
  • Securely attaches to BOLT™ Task Full Face Shield Frame with 5 mounting points
Catalog No. 50-007-68096 Supplier MSC Supplier No. 79945770
The Milwaukee Tool Uncoated Full Face Shield Replacement Lens is designed for use with the BOLT™ Task Full Face Shield Frame, providing protection exceeding ANSI/ISEA Z87.1 (+) - 2020 and CSA Z94.3 standards when used together.
  • Contoured fit
  • For use with BOLT™ Task Full Face Shield Frame
  • Includes 1 clear uncoated replacement shield
  • Offers 180° field of view
  • Compatible with safety glasses
  • Tool free shield change
  • Securely attaches to BOLT™ Task Full Face Shield Frame with 5 mounting points