Learn More
MSC Milwaukee Tool General Purpose Gloves, Small, Polyester, Tricot-Lined, 1 Pair
Catalog No. 5000770182
Encompass
These general purpose gloves are designed for light material handling applications.
  • Small size (numeric size 7)
  • Polyester back material and primary material
  • Tricot lining for comfort
  • Hook & loop cuff style with a 1 inch cuff length
  • Terrycloth thumb brow wipe
  • Full finger coverage
  • Palms and fingertips reinforced with Armortex for ultimate durability and grip
  • Synthetic leather padded palm
  • Maximum operating temperature of 105 degrees F
Catalog No. 50-007-70182 Supplier MSC Supplier No. 10883536
These general purpose gloves are designed for light material handling applications.
  • Small size (numeric size 7)
  • Polyester back material and primary material
  • Tricot lining for comfort
  • Hook & loop cuff style with a 1 inch cuff length
  • Terrycloth thumb brow wipe
  • Full finger coverage
  • Palms and fingertips reinforced with Armortex for ultimate durability and grip
  • Synthetic leather padded palm
  • Maximum operating temperature of 105 degrees F