Learn More
MSC Milwaukee Tool Gloves, Cut A4, Nitrile Dipped, Small, 1 ea
Catalog No. 5000769816
Encompass
These High-Visibility ANSI/ISEA 105-2016 Cut Level 4 Polyurethane Dipped Gloves offer superior fit and protection. They feature a reinforced nitrile coating between the thumb and index finger for enhanced durability and SMARTSWIPE™ touchscreen compatible fingertips. Crafted with high-performance polyethylene, they provide cut protection in a lightweight, breathable design for comfortable all-day wear.
  • High-visibility, cut-resistant design with black palms
  • SMARTSWIPE™ touchscreen compatible fingertips
  • Reinforced nitrile coating between thumb and index finger for enhanced durability
  • High-performance polyethylene knit provides cut protection
  • Lightweight and breathable for comfortable all-day wear
  • Color ID bands for easy cut level identification
Catalog No. 50-007-69816 Supplier MSC Supplier No. 10883916
These High-Visibility ANSI/ISEA 105-2016 Cut Level 4 Polyurethane Dipped Gloves offer superior fit and protection. They feature a reinforced nitrile coating between the thumb and index finger for enhanced durability and SMARTSWIPE™ touchscreen compatible fingertips. Crafted with high-performance polyethylene, they provide cut protection in a lightweight, breathable design for comfortable all-day wear.
  • High-visibility, cut-resistant design with black palms
  • SMARTSWIPE™ touchscreen compatible fingertips
  • Reinforced nitrile coating between thumb and index finger for enhanced durability
  • High-performance polyethylene knit provides cut protection
  • Lightweight and breathable for comfortable all-day wear
  • Color ID bands for easy cut level identification