Learn More
MSC Milwaukee Tool Hard Hat, Class E, 4-Point Suspension, 1 ea
Catalog No. 5000781944
Encompass
The Milwaukee Tool Hard Hat is a construction-grade safety helmet designed for demanding work environments. Featuring a front brim, Class E electrical hazard rating, and a 4-point suspension system, it provides robust head protection. It is constructed from High-Density Polyethylene (HDPE) and is ANSI Type I compliant.
  • Front brim style for sun and glare protection
  • Class E rating provides electrical hazard protection
  • Four-Point suspension system for comfort and impact distribution
  • Adjustable quick release suspension for a secure fit
  • Constructed from high-density polyethylene (HDPE)
  • ANSI Type I and Z89.1 Type 1 compliant
  • Universal size to fit head sizes 6-1/2 to 8
Catalog No. 50-007-81944 Supplier MSC Supplier No. 11641941
The Milwaukee Tool Hard Hat is a construction-grade safety helmet designed for demanding work environments. Featuring a front brim, Class E electrical hazard rating, and a 4-point suspension system, it provides robust head protection. It is constructed from High-Density Polyethylene (HDPE) and is ANSI Type I compliant.
  • Front brim style for sun and glare protection
  • Class E rating provides electrical hazard protection
  • Four-Point suspension system for comfort and impact distribution
  • Adjustable quick release suspension for a secure fit
  • Constructed from high-density polyethylene (HDPE)
  • ANSI Type I and Z89.1 Type 1 compliant
  • Universal size to fit head sizes 6-1/2 to 8