Learn More
MSC Milwaukee Tool Hard Hat, Class E, 6-Point Suspension, 1 ea
Catalog No. 5000687057
Encompass
This yellow HDPE construction hard hat features a front brim and a 6-point suspension system, designed for demanding construction environments.
  • Front brim style hard hat
  • Yellow color
  • Quick release and adjustable 6-point suspension
  • Designed for construction applications
  • Constructed from HDPE material
  • ANSI type I and class E rated
  • Non-vented and non-slotted design
  • Cap brim style
  • Fits head sizes 6-1/2 to 8
  • Overall height: 10.50 in.
  • Overall width: 7.00 in.
  • Overall length: 13.00 in.
  • Meets ANSI Z89.1 type 1 standards
Catalog No. 50-006-87057 Supplier MSC Supplier No. 11641958
This yellow HDPE construction hard hat features a front brim and a 6-point suspension system, designed for demanding construction environments.
  • Front brim style hard hat
  • Yellow color
  • Quick release and adjustable 6-point suspension
  • Designed for construction applications
  • Constructed from HDPE material
  • ANSI type I and class E rated
  • Non-vented and non-slotted design
  • Cap brim style
  • Fits head sizes 6-1/2 to 8
  • Overall height: 10.50 in.
  • Overall width: 7.00 in.
  • Overall length: 13.00 in.
  • Meets ANSI Z89.1 type 1 standards