Learn More
MSC Milwaukee Tool Hard Hat, Front Brim, 6-Point, 1 ea
Catalog No. 5000676701
Encompass
This hard hat is designed for construction applications, featuring a front brim and a 6-point suspension system for comfort and safety, and is made from durable HDPE material.
  • Front brim style
  • Blue color
  • Quick release and adjustable suspension
  • Suitable for construction applications
  • Constructed from HDPE material
  • ANSI type I rated
  • Class E electrical protection
  • Non-vented design
  • Non-slotted design
  • Cap brim style
  • Fits head sizes 6-1/2 to 8
  • Meets ANSI Z89.1 type 1 standards
  • Six-point suspension system
  • Universal size
Catalog No. 50-006-76701 Supplier MSC Supplier No. 11642121
This hard hat is designed for construction applications, featuring a front brim and a 6-point suspension system for comfort and safety, and is made from durable HDPE material.
  • Front brim style
  • Blue color
  • Quick release and adjustable suspension
  • Suitable for construction applications
  • Constructed from HDPE material
  • ANSI type I rated
  • Class E electrical protection
  • Non-vented design
  • Non-slotted design
  • Cap brim style
  • Fits head sizes 6-1/2 to 8
  • Meets ANSI Z89.1 type 1 standards
  • Six-point suspension system
  • Universal size