Learn More
MSC Milwaukee Tool Hard Hat, Full Brim, Class E, 1 ea
Catalog No. 5000787178
Encompass
This hard hat is designed for construction applications, featuring a full brim and a 6-point suspension system. It is gray in color and made from HDPE material. The hard hat is ANSI Type I and Class E rated.
  • Full brim style
  • Gray color
  • Quick release; adjustable
  • Construction application
  • HDPE material
  • ANSI Type I
  • Class E rating
  • Not vented
  • Not slotted
  • No graphics
  • Fits head size 6-1/2 to 8
  • Overall height 10.5 in
  • Overall width 7.0 in
  • Overall length 13.0 in
  • Meets ANSI Z89.1 Type 1 standards
  • Universal size
  • Six-point suspension
Catalog No. 50-007-87178 Supplier MSC Supplier No. 11642287
This hard hat is designed for construction applications, featuring a full brim and a 6-point suspension system. It is gray in color and made from HDPE material. The hard hat is ANSI Type I and Class E rated.
  • Full brim style
  • Gray color
  • Quick release; adjustable
  • Construction application
  • HDPE material
  • ANSI Type I
  • Class E rating
  • Not vented
  • Not slotted
  • No graphics
  • Fits head size 6-1/2 to 8
  • Overall height 10.5 in
  • Overall width 7.0 in
  • Overall length 13.0 in
  • Meets ANSI Z89.1 Type 1 standards
  • Universal size
  • Six-point suspension