Learn More
MSC Milwaukee Tool Hard Hat, Full Brim, Class E, 1 ea
Catalog No. 5000787129
Encompass
This full brim hard hat is constructed from high-density polyethylene, designed for construction applications. It features a 6-point suspension system for comfort and safety, and meets ANSI Type I and Class E standards.
  • Constructed from high-density polyethylene
  • Features a full brim style
  • Equipped with a quick release, adjustable suspension system
  • Suitable for construction applications
  • ANSI Type I and Class E rated
  • Not vented or slotted
  • Fits head sizes from 6-1/2 to 8
  • Complies with ANSI Z89.1 Type 1 standards
  • Includes a 6-point suspension
Catalog No. 50-007-87129 Supplier MSC Supplier No. 11642097
This full brim hard hat is constructed from high-density polyethylene, designed for construction applications. It features a 6-point suspension system for comfort and safety, and meets ANSI Type I and Class E standards.
  • Constructed from high-density polyethylene
  • Features a full brim style
  • Equipped with a quick release, adjustable suspension system
  • Suitable for construction applications
  • ANSI Type I and Class E rated
  • Not vented or slotted
  • Fits head sizes from 6-1/2 to 8
  • Complies with ANSI Z89.1 Type 1 standards
  • Includes a 6-point suspension