Learn More
MSC Milwaukee Tool Hard Hat, Full Brim, Class E, 1 ea
Catalog No. 5000782337
Encompass
This hard hat is designed for construction applications, featuring a full brim, Class E protection, and a 6-point suspension system for optimal safety and comfort.
  • Full brim style
  • Red color
  • Quick release and adjustable fit
  • Made from HDPE material
  • ANSI Type I, Class E rated
  • Fits head sizes 6-1/2 to 8
  • Overall height 10.5 in
  • Overall width 7.0 in
  • Overall length 13.0 in
  • Meets ANSI Z89.1 Type 1 standards
  • Universal size
  • Six-point suspension
Catalog No. 50-007-82337 Supplier MSC Supplier No. 11642055
This hard hat is designed for construction applications, featuring a full brim, Class E protection, and a 6-point suspension system for optimal safety and comfort.
  • Full brim style
  • Red color
  • Quick release and adjustable fit
  • Made from HDPE material
  • ANSI Type I, Class E rated
  • Fits head sizes 6-1/2 to 8
  • Overall height 10.5 in
  • Overall width 7.0 in
  • Overall length 13.0 in
  • Meets ANSI Z89.1 Type 1 standards
  • Universal size
  • Six-point suspension