Learn More
MSC Milwaukee Tool Hard Hat Full Brim Class E, 1 ea
Catalog No. 5000687055
Encompass
This product is a blue hard hat made of HDPE, designed for construction applications. It features a full brim and a 4-point suspension system. It is ANSI Type I and Class E rated, and it is not vented or slotted. The hard hat fits head sizes from 6-1/2 to 8.
  • Full brim style
  • Blue color
  • Quick release; adjustable fit
  • Designed for construction applications
  • Made of HDPE material
  • ANSI Type I
  • Not vented
  • Not slotted
  • No graphics
  • Fits head sizes 6-1/2 to 8
  • Overall height: 10.20 in.
  • Overall width: 6.00 in.
  • Overall length: 12.50 in.
  • Meets ANSI Z89.1 Type 1 standards
  • Universal size
  • Four-point suspension
  • Class E rating
Catalog No. 50-006-87055 Supplier MSC Supplier No. 11641792
This product is a blue hard hat made of HDPE, designed for construction applications. It features a full brim and a 4-point suspension system. It is ANSI Type I and Class E rated, and it is not vented or slotted. The hard hat fits head sizes from 6-1/2 to 8.
  • Full brim style
  • Blue color
  • Quick release; adjustable fit
  • Designed for construction applications
  • Made of HDPE material
  • ANSI Type I
  • Not vented
  • Not slotted
  • No graphics
  • Fits head sizes 6-1/2 to 8
  • Overall height: 10.20 in.
  • Overall width: 6.00 in.
  • Overall length: 12.50 in.
  • Meets ANSI Z89.1 Type 1 standards
  • Universal size
  • Four-point suspension
  • Class E rating