Learn More
MSC Milwaukee Tool Hard Hat, Full Brim, Class E, 1 ea
Catalog No. 5000796442
Encompass
This full brim hard hat is designed for construction applications, providing robust head protection with an ANSI Type I and Class E rating.
  • Made of HDPE
  • Black color
  • Has a 4-point suspension
  • Quick release, adjustable adjustment type
  • Fits head sizes 6-1/2 to 8
  • Conforms to ANSI Z89.1 Type 1 standards
  • Universal size
Catalog No. 50-007-96442 Supplier MSC Supplier No. 11641842
This full brim hard hat is designed for construction applications, providing robust head protection with an ANSI Type I and Class E rating.
  • Made of HDPE
  • Black color
  • Has a 4-point suspension
  • Quick release, adjustable adjustment type
  • Fits head sizes 6-1/2 to 8
  • Conforms to ANSI Z89.1 Type 1 standards
  • Universal size