Learn More
MSC Milwaukee Tool Heated Base Layer Shirt, 2X-Large, 1 ea
Catalog No. 5000686913
Encompass
This heated midweight base layer shirt is designed to provide warmth and comfort in cold conditions. It features carbon fiber heating elements to distribute heat to the chest and back, powered by a USB rechargeable battery.
  • Quick-heat function reaches selected temperature 3X faster
  • One-touch LED controller with 2 heat settings
  • Up to 3-hours of run-time with battery pack
  • Heating zones: chest and upper back
  • Washer and dryer safe
  • Midweight construction keeps body warm and dry
  • Fleece-lined arms provide total body warmth
  • Raglan sleeves and seamless shoulders provide improved mobility and comfort
  • Pilling resistant fabric extends product life
Catalog No. 50-006-86913 Supplier MSC Supplier No. 14845242
This heated midweight base layer shirt is designed to provide warmth and comfort in cold conditions. It features carbon fiber heating elements to distribute heat to the chest and back, powered by a USB rechargeable battery.
  • Quick-heat function reaches selected temperature 3X faster
  • One-touch LED controller with 2 heat settings
  • Up to 3-hours of run-time with battery pack
  • Heating zones: chest and upper back
  • Washer and dryer safe
  • Midweight construction keeps body warm and dry
  • Fleece-lined arms provide total body warmth
  • Raglan sleeves and seamless shoulders provide improved mobility and comfort
  • Pilling resistant fabric extends product life