Learn More
MSC Milwaukee Tool Heated Jacket, Women's, M12 AXIS, Insulated, 1 ea
Catalog No. 5000686208
Encompass
This women's heated jacket offers lightweight durability, powered by REDLITHIUM™ Battery Technology. It features an advanced heat system with carbon fiber elements and Quick-Heat functionality, distributing heat faster to the chest, back, and shoulders. Battery pass-thru allows versatile placement.
  • Advanced heat system for 3X faster heat-up
  • Battery pass-thru for versatile placement
  • Up to 12 hours run-time on low setting
  • Three heat zones: chest, back, shoulders
  • One-touch LED controller with three heat settings
  • Five total pockets for maximized storage
  • AXIS™ ripstop polyester and high-wear zone reinforcement
  • Wind and water resistant with DWR coating
  • Polyester insulation, no bulk
  • Adjustable waist
  • Washer and dryer safe
Catalog No. 50-006-86208 Supplier MSC Supplier No. 01101344
This women's heated jacket offers lightweight durability, powered by REDLITHIUM™ Battery Technology. It features an advanced heat system with carbon fiber elements and Quick-Heat functionality, distributing heat faster to the chest, back, and shoulders. Battery pass-thru allows versatile placement.
  • Advanced heat system for 3X faster heat-up
  • Battery pass-thru for versatile placement
  • Up to 12 hours run-time on low setting
  • Three heat zones: chest, back, shoulders
  • One-touch LED controller with three heat settings
  • Five total pockets for maximized storage
  • AXIS™ ripstop polyester and high-wear zone reinforcement
  • Wind and water resistant with DWR coating
  • Polyester insulation, no bulk
  • Adjustable waist
  • Washer and dryer safe