Learn More
MSC Milwaukee Tool High Visibility Vest, ANSI Class 3, 1 ea
Catalog No. 5000789346
Encompass
This high-visibility vest is designed for public safety, meeting ANSI Class 3 standards for enhanced visibility. It features a durable mesh construction and multiple pockets for practical utility.
  • Available in large and X-large sizes
  • Lime color for high visibility
  • Zipper closure for secure fit
  • Features 10 pockets for storage
  • Mesh material composition
  • Equipped with 3 silver horizontal stripes, 2 in wide
  • ANSI Class 3 certified
Catalog No. 50-007-89346 Supplier MSC Supplier No. 20882551
This high-visibility vest is designed for public safety, meeting ANSI Class 3 standards for enhanced visibility. It features a durable mesh construction and multiple pockets for practical utility.
  • Available in large and X-large sizes
  • Lime color for high visibility
  • Zipper closure for secure fit
  • Features 10 pockets for storage
  • Mesh material composition
  • Equipped with 3 silver horizontal stripes, 2 in wide
  • ANSI Class 3 certified