Learn More
MSC Milwaukee Tool Inspection Camera, 18 mm Scope, 75 ft Viewing Distance, 7 in Display
Catalog No. 5000531192
Encompass
The Milwaukee Tool inspection camera is designed for detailed visual inspections in various applications. It features an 18 mm scope, a maximum viewing distance of 75 ft, and a 7 in display, providing clear visuals for identifying issues in hard-to-reach areas.
  • Inspection camera type
  • 18 mm scope diameter
  • 75 ft maximum viewing distance
  • 7 in display size
  • Adjustable focus
  • LED bulb type
  • Alkaline battery chemistry
  • 12V battery size
  • 75 ft cable length
  • 0.1250 in cable diameter
  • LCD display type
  • Packout compatibility
Catalog No. 50-005-31192 Supplier MSC Supplier No. 65600769
The Milwaukee Tool inspection camera is designed for detailed visual inspections in various applications. It features an 18 mm scope, a maximum viewing distance of 75 ft, and a 7 in display, providing clear visuals for identifying issues in hard-to-reach areas.
  • Inspection camera type
  • 18 mm scope diameter
  • 75 ft maximum viewing distance
  • 7 in display size
  • Adjustable focus
  • LED bulb type
  • Alkaline battery chemistry
  • 12V battery size
  • 75 ft cable length
  • 0.1250 in cable diameter
  • LCD display type
  • Packout compatibility