Learn More
MSC Milwaukee Tool Inspection Camera Cable, 10 ft, M12 Drain Camera Compatible, Waterproof
Catalog No. 5000531193
Encompass
This accessory cable for inspection cameras and borescopes is designed for use with compatible drain cameras. The cable measures 10 ft in length and is waterproof. It includes a 10 mm borescope camera cable with an HD camera that delivers clear images and allows for easy access in confined environments.
  • Accessory cable for inspection cameras and borescopes.
  • Compatible with drain cameras.
  • Waterproof design.
  • Measures 10 ft in length.
  • Includes 10 mm borescope camera cable.
  • HD camera delivers clear images.
  • Easy access in confined environments.
Catalog No. 50-005-31193 Supplier MSC Supplier No. 65601403
This accessory cable for inspection cameras and borescopes is designed for use with compatible drain cameras. The cable measures 10 ft in length and is waterproof. It includes a 10 mm borescope camera cable with an HD camera that delivers clear images and allows for easy access in confined environments.
  • Accessory cable for inspection cameras and borescopes.
  • Compatible with drain cameras.
  • Waterproof design.
  • Measures 10 ft in length.
  • Includes 10 mm borescope camera cable.
  • HD camera delivers clear images.
  • Easy access in confined environments.