Learn More
MSC Milwaukee Tool M18 FUEL 2.5 gal. Wet/Dry Vacuum, 1 Kit
Catalog No. 5000788253
Encompass
The Milwaukee Tool M18 FUEL™ 2.5 gal. Wet/Dry Vacuum with PACKOUT™ Compatibility is a battery-powered solution designed for high-speed applications. It features a robust polycarbonate tank and efficient HEPA filtration.
  • Battery powered operation
  • HEPA filtration for efficient dust collection
  • Collection capacity: 2.5 gal.
  • Bagless vacuum collection
  • Polycarbonate tank material
  • Maximum air flow of 50
  • Hose length: 2 ft
  • Vacuum hose fitting size: 1-1/4 inch
  • Includes a power tool adapter
  • Includes a long reach flexible micro hose set
  • Includes a 6 ft flexible hose, crevice tool, and utility nozzle
  • Compatible with PACKOUT storage systems
Catalog No. 50-007-88253 Supplier MSC Supplier No. 68737022
The Milwaukee Tool M18 FUEL™ 2.5 gal. Wet/Dry Vacuum with PACKOUT™ Compatibility is a battery-powered solution designed for high-speed applications. It features a robust polycarbonate tank and efficient HEPA filtration.
  • Battery powered operation
  • HEPA filtration for efficient dust collection
  • Collection capacity: 2.5 gal.
  • Bagless vacuum collection
  • Polycarbonate tank material
  • Maximum air flow of 50
  • Hose length: 2 ft
  • Vacuum hose fitting size: 1-1/4 inch
  • Includes a power tool adapter
  • Includes a long reach flexible micro hose set
  • Includes a 6 ft flexible hose, crevice tool, and utility nozzle
  • Compatible with PACKOUT storage systems