Learn More
MSC Milwaukee Tool Nitrile Dipped Gloves, Cut A5, Large, 1 Pair
Catalog No. 5000802885
Encompass
The ANSI/ISEA 105-2016 Cut Level 5 Nitrile Dipped Gloves are engineered for enhanced durability and superior protection. They feature a reinforced nitrile coating between the thumb and index finger, specifically designed to withstand wear in high-stress areas.
  • Reinforced nitrile coating between thumb and index finger
  • Nitrile coated for better grip
  • SMARTSWIPE™ touchscreen compatible fingertips
  • Breathable glove design
  • ANSI/ISEA 105-2016 Cut Level 5
  • EN388 - 4X43E
  • Nylon/HPPE blend liner
  • Color-coded wristband and logo for easy cut level identification
Catalog No. 50-008-02885 Supplier MSC Supplier No. 77169480
The ANSI/ISEA 105-2016 Cut Level 5 Nitrile Dipped Gloves are engineered for enhanced durability and superior protection. They feature a reinforced nitrile coating between the thumb and index finger, specifically designed to withstand wear in high-stress areas.
  • Reinforced nitrile coating between thumb and index finger
  • Nitrile coated for better grip
  • SMARTSWIPE™ touchscreen compatible fingertips
  • Breathable glove design
  • ANSI/ISEA 105-2016 Cut Level 5
  • EN388 - 4X43E
  • Nylon/HPPE blend liner
  • Color-coded wristband and logo for easy cut level identification