Learn More
MSC Milwaukee Tool Power Bit, SHOCKWAVE Impact Duty, 3-1/2 in, TORX T20
Catalog No. 5000513731
Encompass
Our SHOCKWAVE Impact Duty 3-1/2 in T20 Driver Bits provide the Ultimate Fit for a superior driving experience. The optimized SHOCKZONE shanks are customized per tip type to absorb peak torque and prevent breaking. The long-lasting Wear Guard Tip provides increased wear resistance, protecting the fit over the life of the bit. The customized tip geometry leads to less stripping of screws on the job site.
  • Ultimate fit
  • Most durable, SHOCKZONE shank absorbs peak torque
  • Long lasting tip, Wear Guard
  • Customized tip geometry, less stripping
  • SHOCKZONE shanks are optimized per tip type to prevent breaking
  • Custom heat treat for longer bit life
  • Laser etched for easy size identification
  • Designed for use with impact drivers and drill drivers
Catalog No. 50-005-13731 Supplier MSC Supplier No. 32804601
Our SHOCKWAVE Impact Duty 3-1/2 in T20 Driver Bits provide the Ultimate Fit for a superior driving experience. The optimized SHOCKZONE shanks are customized per tip type to absorb peak torque and prevent breaking. The long-lasting Wear Guard Tip provides increased wear resistance, protecting the fit over the life of the bit. The customized tip geometry leads to less stripping of screws on the job site.
  • Ultimate fit
  • Most durable, SHOCKZONE shank absorbs peak torque
  • Long lasting tip, Wear Guard
  • Customized tip geometry, less stripping
  • SHOCKZONE shanks are optimized per tip type to prevent breaking
  • Custom heat treat for longer bit life
  • Laser etched for easy size identification
  • Designed for use with impact drivers and drill drivers