Learn More
MSC Milwaukee Tool Power Bit, SHOCKWAVE Impact Duty, TORX T15, 2 in
Supplier: MSC 32802647
The Milwaukee Tool SHOCKWAVE Impact Duty 2-inch TORX T15 Power Bit provides ultimate fit for a superior driving experience. Its optimized SHOCKZONE shanks absorb peak torque to prevent breaking, and the long-lasting Wear Guard Tip increases wear resistance, protecting the fit over the life of the bit. The customized tip geometry reduces screw stripping on the job site.
- Ultimate fit
- Most durable, SHOCKZONE shank absorbs peak torque
- Long lasting tip, Wear Guard
- Customized tip geometry, less stripping
- SHOCKZONE shanks are optimized per tip type to prevent breaking
- Custom heat treat for longer bit life
- Laser etched for easy size identification
- Designed for use with impact drivers and drill drivers