Learn More
MSC Milwaukee Tool Safety Glasses, Anti-Fog, Gray Lenses, Half-Framed, 1 ea
Catalog No. 5000770028
Encompass
These safety glasses are designed with anti-fog and scratch-resistant gray plastic lenses, featuring a half-framed, wrap-around design for enhanced protection and visibility.
  • Anti-fog coatings improve visibility in hot and humid conditions
  • Scratch-resistant lenses
  • Gray plastic lenses
  • Half-framed, wrap-around design
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Fixed temple type
Catalog No. 50-007-70028 Supplier MSC Supplier No. 10882033
These safety glasses are designed with anti-fog and scratch-resistant gray plastic lenses, featuring a half-framed, wrap-around design for enhanced protection and visibility.
  • Anti-fog coatings improve visibility in hot and humid conditions
  • Scratch-resistant lenses
  • Gray plastic lenses
  • Half-framed, wrap-around design
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Fixed temple type