Learn More
MSC Milwaukee Tool Safety Vest, AR/FR Class 2, 1 ea
Catalog No. 5000792121
Encompass
This AR/FR Cat. 1 Class 2 High Visibility Yellow Safety Vest is engineered for arc flash and flame resistance, providing essential protection and visibility in hazardous work environments. It features durable construction and multiple pockets for convenient storage.
  • Ten pockets for ample storage, including mic tabs, chest pockets, and pen pockets
  • Fall protection pass through for harness compatibility
  • Hook and loop closure for secure fastening
  • Internal size adjustment for a customized fit
  • Constructed from WESTEX® DH fabric for durability
  • Arc rating of ATPV 5.7 CAL./cm²
  • Meets ANSI/ISEA 107-15 Type R Class 2 standard
  • UL Certified to ASTM F1506 and NFPA 2112
Catalog No. 50-007-92121 Supplier MSC Supplier No. 82853904
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This AR/FR Cat. 1 Class 2 High Visibility Yellow Safety Vest is engineered for arc flash and flame resistance, providing essential protection and visibility in hazardous work environments. It features durable construction and multiple pockets for convenient storage.
  • Ten pockets for ample storage, including mic tabs, chest pockets, and pen pockets
  • Fall protection pass through for harness compatibility
  • Hook and loop closure for secure fastening
  • Internal size adjustment for a customized fit
  • Constructed from WESTEX® DH fabric for durability
  • Arc rating of ATPV 5.7 CAL./cm²
  • Meets ANSI/ISEA 107-15 Type R Class 2 standard
  • UL Certified to ASTM F1506 and NFPA 2112