Learn More
MSC Milwaukee Tool Safety Vest, Class 2, Yellow, 4X/5X, 1 ea
Catalog No. 5000796673
Encompass
Our ANSI Type R Class 2 High Visibility Safety Vests are designed to carry more. These safety vests feature a durable pass-through slit for fall protection gear and a zipper closure for jobsite security. They are recommended for environments with traffic traveling under 50 mph or around heavy machinery, including construction and road work.
  • Ten pockets for ample storage, including a clear ID holder
  • Secure zipper closure for ultimate jobsite security
  • Fall protection pass through for harness compatibility
  • Reinforced pen pockets with 600D tear resistant material
  • Durable dual mic tabs
  • Internal pocket for additional storage
  • ANSI107-15 Class 2 Type R compliant
Catalog No. 50-007-96673 Supplier MSC Supplier No. 10882918
Our ANSI Type R Class 2 High Visibility Safety Vests are designed to carry more. These safety vests feature a durable pass-through slit for fall protection gear and a zipper closure for jobsite security. They are recommended for environments with traffic traveling under 50 mph or around heavy machinery, including construction and road work.
  • Ten pockets for ample storage, including a clear ID holder
  • Secure zipper closure for ultimate jobsite security
  • Fall protection pass through for harness compatibility
  • Reinforced pen pockets with 600D tear resistant material
  • Durable dual mic tabs
  • Internal pocket for additional storage
  • ANSI107-15 Class 2 Type R compliant