Learn More
MSC Milwaukee Tool Self-Adhering Tape, 12 ft, Fiberglass Reinforced, 1 ea
Catalog No. 5000796248
Encompass
This self-adhering tape is designed to create a secure attachment point on most tools without a lanyard hole. It adheres only to itself, leaving no residue on tools, and is fiberglass reinforced for high strength. Compatible with a wide range of tools, it is intended for use with D-Ring Web Attachments or Quick-Connect Accessories.
  • Creates a secure attachment point for tethering
  • Self-adhering, uses no adhesives
  • Leaves no residue on tools
  • Fiberglass reinforced for high strength
  • Compatible with a wide range of tools
  • For use with D-Ring web attachments
  • Limited lifetime warranty
  • Third-party tested at 2:1 safety factor
Catalog No. 50-007-96248 Supplier MSC Supplier No. 19942002
This self-adhering tape is designed to create a secure attachment point on most tools without a lanyard hole. It adheres only to itself, leaving no residue on tools, and is fiberglass reinforced for high strength. Compatible with a wide range of tools, it is intended for use with D-Ring Web Attachments or Quick-Connect Accessories.
  • Creates a secure attachment point for tethering
  • Self-adhering, uses no adhesives
  • Leaves no residue on tools
  • Fiberglass reinforced for high strength
  • Compatible with a wide range of tools
  • For use with D-Ring web attachments
  • Limited lifetime warranty
  • Third-party tested at 2:1 safety factor