Learn More
MSC Milwaukee Tool Sprayer Nozzles, Adjustable, Fan, M18 Sprayer, 2 Pack
Catalog No. 5000792109
Encompass
Milwaukee Tool Sprayer Nozzles are designed to provide versatile spray patterns for M18 Backpack Sprayers. These nozzles offer options for various applications, including targeted control and wide area coverage.
  • Adjustable nozzle allows for cone shape to straight stream to mist spray patterns
  • Adjustable nozzle has small diameter outlet for more control in targeted applications
  • Fan nozzle delivers a wide, flat spray pattern
Catalog No. 50-007-92109 Supplier MSC Supplier No. 81095333
Milwaukee Tool Sprayer Nozzles are designed to provide versatile spray patterns for M18 Backpack Sprayers. These nozzles offer options for various applications, including targeted control and wide area coverage.
  • Adjustable nozzle allows for cone shape to straight stream to mist spray patterns
  • Adjustable nozzle has small diameter outlet for more control in targeted applications
  • Fan nozzle delivers a wide, flat spray pattern