Learn More
MSC Milwaukee Tool Tool Case, Customizable Insert, 75 lb Capacity, Impact Resistant Polymer
Catalog No. 5000516753
Encompass
The PACKOUT™ Modular Storage System offers a durable and versatile storage solution. This tool case features an impact-resistant molded base that integrates with other PACKOUT™ components. Constructed from impact-resistant polymer with a metal-reinforced locking point, it is designed for harsh job site environments. It includes an IP65-rated weather seal to protect contents and a customizable foam insert for secure tool storage.
  • 75 lb weight capacity
  • Heavy duty latches
  • Impact resistant polymer construction
  • Customizable foam insert included
  • IP65 rated weather seal
  • Metal reinforced locking point
  • Mounting location for tool and equipment tracker
  • Part of the modular storage system
  • Reinforced hinge
  • Modular connectivity with all components
  • Limited lifetime warranty
Catalog No. 50-005-16753 Supplier MSC Supplier No. 12765038
The PACKOUT™ Modular Storage System offers a durable and versatile storage solution. This tool case features an impact-resistant molded base that integrates with other PACKOUT™ components. Constructed from impact-resistant polymer with a metal-reinforced locking point, it is designed for harsh job site environments. It includes an IP65-rated weather seal to protect contents and a customizable foam insert for secure tool storage.
  • 75 lb weight capacity
  • Heavy duty latches
  • Impact resistant polymer construction
  • Customizable foam insert included
  • IP65 rated weather seal
  • Metal reinforced locking point
  • Mounting location for tool and equipment tracker
  • Part of the modular storage system
  • Reinforced hinge
  • Modular connectivity with all components
  • Limited lifetime warranty