Learn More
Amsbio LLC Mimetix 12 well plate for 3D cell culture (loose discs, retaining rings), sterile
Catalog No. 502041588
Encompass
Mimetix scaffolds mimic the extracellular matrix and provide an ideal architectural environment to support the growth of cells in 3D. They are created by electrospinning medical-grade polymer poly(L-lactide) (PLLA) into microfibers, which are highly consistent with regard to fibre diameter and pore size, resulting in excellent reproducibility of cell-based assays.
Catalog No. 50-204-1588 Supplier Amsbio LLC Supplier No. AMS.TECL0031X
Mimetix scaffolds mimic the extracellular matrix and provide an ideal architectural environment to support the growth of cells in 3D. They are created by electrospinning medical-grade polymer poly(L-lactide) (PLLA) into microfibers, which are highly consistent with regard to fibre diameter and pore size, resulting in excellent reproducibility of cell-based assays.