Learn More
Braintree Scientific Mimicky Mouse Model
SDP
Catalog No. 501955093
Encompass
Mimicky Mouse Mimicky assists in the training of mouse handling techniques and procedures. By using this mouse type simulator, you contribute to the realization of the philosophy of the 3 R's. By using alternative training and treatment before use, more humane principles, can be achieved. This is a simulator that mimics the experimental animals (mice) Mimicky (mimicking) the feel when dealing with biometric mice. Handling Restraint Physical assessment Oral administration Intravenous administration Mimicky Mouse Body:Approx. 70 (L) x 39 (H) x 31 (W) mm 25gMimicky Tail:Approx. 130 mm (exposed section approx. 110 mm) 3gWeight: approx. 28g **Sold for research purposes only and not for human use**
Catalog No. 50-195-5093 Supplier Braintree Scientific Supplier No. MIMICKY1
Mimicky Mouse Mimicky assists in the training of mouse handling techniques and procedures. By using this mouse type simulator, you contribute to the realization of the philosophy of the 3 R's. By using alternative training and treatment before use, more humane principles, can be achieved. This is a simulator that mimics the experimental animals (mice) Mimicky (mimicking) the feel when dealing with biometric mice. Handling Restraint Physical assessment Oral administration Intravenous administration Mimicky Mouse Body:Approx. 70 (L) x 39 (H) x 31 (W) mm 25gMimicky Tail:Approx. 130 mm (exposed section approx. 110 mm) 3gWeight: approx. 28g **Sold for research purposes only and not for human use**