Learn More
YSI 3P LifeScience 1531 Glucose Standard, 900 mg/dL (9 g/L)
Catalog No. 50200107
Encompass
900 mg/dL (9 g/L, 50 mmol/L) Glucose standard solution for use with YSI 2300, 2700 and 2900 Series Analyzers.
Catalog No. 50-200-107 Supplier YSI 3P LifeScience Supplier No. 015053
900 mg/dL (9 g/L, 50 mmol/L) Glucose standard solution for use with YSI 2300, 2700 and 2900 Series Analyzers.