Learn More
Grainger Ideal Tridon Worm Gear Hose Clamp, 301 Stainless Steel, 11/16 in. to 1 1/2 in. Clamping Diameter, 10/Pk.
Catalog No. NC0678032
Encompass_Preferred
These micro-gear hose clamps are precision-engineered for use in small hose applications, such as emission control, vacuum lines, and fuel hoses. Designed with a slim profile, these clamps allow for installation in confined spaces where standard clamps may not fit. They are constructed from durable 301 stainless steel to provide reliable sealing performance.
  • Clamping diameter range: 11/16 in. to 1-1/2 in. (18 mm to 38 mm)
  • Band width: 5/16 in.
  • Constructed from 301 stainless steel for corrosion resistance
  • Low profile housing suitable for confined areas
  • Torque rating: 10 lbf-in. to 15 lbf-in.
  • 1/4 in. hex head size
  • Package includes 10 clamps
Catalog No. NC0678032 Supplier Grainger Supplier No. 5CZD5
These micro-gear hose clamps are precision-engineered for use in small hose applications, such as emission control, vacuum lines, and fuel hoses. Designed with a slim profile, these clamps allow for installation in confined spaces where standard clamps may not fit. They are constructed from durable 301 stainless steel to provide reliable sealing performance.
  • Clamping diameter range: 11/16 in. to 1-1/2 in. (18 mm to 38 mm)
  • Band width: 5/16 in.
  • Constructed from 301 stainless steel for corrosion resistance
  • Low profile housing suitable for confined areas
  • Torque rating: 10 lbf-in. to 15 lbf-in.
  • 1/4 in. hex head size
  • Package includes 10 clamps