Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermco Products Min-Max Hygrometer Thermometer Internal and External Temperature
Description
- The Min-Max Hygrometer Thermometer is a versatile device for monitoring internal and external temperatures, humidity, and time.
- Features a jumbo display for easy reading.
- Measures internal temperatures from 0°C to 50°C (32°F to 122°F) and external temperatures from -50°C to +70°C (-58°F to +158°F).
- Humidity measurement range from 10% to 99% Rh.
- Stores minimum and maximum readings for quick reference.
- Selectable Celsius or Fahrenheit units.
- Includes a time display with alarm clock function.
- Foldaway stand and mounting keyhole allow flexible placement.
- CE marked for quality assurance and equipped with a low battery indicator.
- Compact design measuring 100 mm (W) x 110 mm (H)
Specifications
Specifications
| Type | Hygrometer |
| Humidity Range | 10 to 99% |
| Temperature Range (Metric) | 0°C to 50°C (Internal), -50°C to +70°C (External) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?