Learn More
Thermco Products Inc MIN/MAX THERMOMETER
Catalog No. NC1174595
Encompass_Preferred
NIST Traceable w/CertificateTri-Display For Current, Min And Max TemperaturesInernal & External Temperature MeasuringProgrammable Low And High Alarm LimitExternal Probe with 3 Meters Cord LengthTemp Range:Internal 0C to 50C ( 32F to 122F)External -50C to 70C (-58F to 158F)Accuracy: 1.0C (1.8F)Resolution: 0.1C/F Switchable10 seconds Display UpdateLow Battery IndicationMagnet Attachment and Hook and Loop Magic Tape Fixing.
Catalog No. NC1174595 Supplier Thermco Products Inc Supplier No. ACC850DIG
NIST Traceable w/CertificateTri-Display For Current, Min And Max TemperaturesInernal & External Temperature MeasuringProgrammable Low And High Alarm LimitExternal Probe with 3 Meters Cord LengthTemp Range:Internal 0C to 50C ( 32F to 122F)External -50C to 70C (-58F to 158F)Accuracy: 1.0C (1.8F)Resolution: 0.1C/F Switchable10 seconds Display UpdateLow Battery IndicationMagnet Attachment and Hook and Loop Magic Tape Fixing.