Learn More
Ace Glass, Inc. 40mL complete mineral oil bubbler, 7mm inlet tube, nylon bushing with FETFE O-ring
SDP
Catalog No. 509558583
Encompass_Preferred
40mL mineral oil bubbler with a tapered vessel bottom. Bubbler is supplied with a 7mm inner tube with a pulled tip, and a nylon bushing with a FETFE o-ring.
Catalog No. 50-955-8583 Supplier Ace Glass, Inc. Supplier No. 876214
40mL mineral oil bubbler with a tapered vessel bottom. Bubbler is supplied with a 7mm inner tube with a pulled tip, and a nylon bushing with a FETFE o-ring.