Learn More
Invent Biotechnologies Inc Minute Plasma Membrane Protein Isolation Kit, 50 preps, cultured mammalian cells or tissues, 1-50 million cells/sample, 10-500 μg/sample, -20 °C
Supplier:  Invent Biotechnologies Inc SM005
Encompass
The Minute Plasma Membrane Protein Isolation Kit provides an efficient method for isolating native total membrane proteins and plasma membrane proteins from cultured mammalian cells or tissues. This kit simplifies the process by utilizing optimized buffers that are both detergent and EDTA-free, alongside protein extraction filter cartridges to help isolate cell components like nuclei, cytosol, organelles, and plasma membrane efficiently within 45 minutes, without traditional homogenization. Suitable for various applications including SDS-PAGE and immunoblottings, this kit accommodates a wide range of cell numbers.
Catalog No. NC0280257