Learn More
Aviva Systems Biology Corp RIOX2 Antibody - N-terminal region (ARP39721_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39721_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MINA. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MINA. Alternate names: MDIG, NO52, MINA53ApplicationWB, IHC. Species reactivity: Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57151