Learn More
MSC Snap-On Air Vacuum, Plastic, 5.00 in Hg Static Pull, 12 in Hose
Catalog No. 502837517
Encompass
The Snap-On Special Products Division MINA-VAC Air Operated Vacuum is an air-operated vacuum manufactured in the USA. It features a one-piece housing with molded-in metal 1/4 NPT threads, offering 25% more suction than previous models with a 5.00 in Hg static pull. It includes a 12-inch clear flexible extension hose and can remove objects up to 3/4 inch in diameter.
  • One-piece housing with molded-in metal 1/4 NPT threads
  • 25% more suction than previous model
  • 5.00 in Hg static pull
  • 12-inch clear flexible extension hose
  • Removes objects as large as 3/4 inch in diameter
  • Built-in chip deflector minimizes bag damage and increases bag life
  • Trigger can lock in place for constant suction
  • Removable bag-clip for easy emptying of debris
  • Manufactured in the USA
Catalog No. 50-283-7517 Supplier MSC Supplier No. 72008105
The Snap-On Special Products Division MINA-VAC Air Operated Vacuum is an air-operated vacuum manufactured in the USA. It features a one-piece housing with molded-in metal 1/4 NPT threads, offering 25% more suction than previous models with a 5.00 in Hg static pull. It includes a 12-inch clear flexible extension hose and can remove objects up to 3/4 inch in diameter.
  • One-piece housing with molded-in metal 1/4 NPT threads
  • 25% more suction than previous model
  • 5.00 in Hg static pull
  • 12-inch clear flexible extension hose
  • Removes objects as large as 3/4 inch in diameter
  • Built-in chip deflector minimizes bag damage and increases bag life
  • Trigger can lock in place for constant suction
  • Removable bag-clip for easy emptying of debris
  • Manufactured in the USA