Learn More
Selleck Chemical LLC Minaprine dihydrochloride
SDP
Catalog No. 5000003076
Encompass_Preferred
Minaprine dihydrochloride is hydrochloride salt form of Minaprine which acts as a reversible inhibitor of MAO-A in rats and weakly inhibit acetylcholinesterase in rat brain (striatum) homogenates
Catalog No. 50-000-03076 Supplier Selleck Chemical LLC Supplier No. S566325MG
Minaprine dihydrochloride is hydrochloride salt form of Minaprine which acts as a reversible inhibitor of MAO-A in rats and weakly inhibit acetylcholinesterase in rat brain (striatum) homogenates