Learn More
Cayman Chemical Minaprine (hydrochloride), Purity: ≥98%, CAS: 25905-77-5, 250 mg
SDP
Catalog No. 502944251
Encompass_Preferred
An antidepressant; reduces immobility time in the forced swim test in rats at 5 mg/kg for nine days; induces convulsions in rats at 40 mg/kg
Catalog No. 50-294-4251 Supplier Cayman Chemical Supplier No. 41710250 MG
An antidepressant; reduces immobility time in the forced swim test in rats at 5 mg/kg for nine days; induces convulsions in rats at 40 mg/kg