Learn More
Medchemexpress LLC MIND4-19 | 129544-85-0 | 99.9% | 373.47 | 5 MG
SDP
Catalog No. 5000361194
Encompass_Preferred
MIND4-19 is a potent SIRT2 inhibitor with an IC50 value of 7.0 μM. This compound is intended for use in researching Huntington's disease.
  • Solid, white to off-white appearance
  • Soluble in DMSO for in vitro applications
  • Can be dissolved using various co-solvents for in vivo protocols
  • Targeted for SIRT2 inhibition
Catalog No. 50-003-61194 Supplier Medchemexpress LLC Supplier No. HY1472335MG
MIND4-19 is a potent SIRT2 inhibitor with an IC50 value of 7.0 μM. This compound is intended for use in researching Huntington's disease.
  • Solid, white to off-white appearance
  • Soluble in DMSO for in vitro applications
  • Can be dissolved using various co-solvents for in vivo protocols
  • Targeted for SIRT2 inhibition