Learn More
Medchemexpress LLC MIND4-19 | 129544-85-0 | 99.93% | 373.47 | 100 MG
SDP
Catalog No. 5000360557
Encompass_Preferred
MIND4-19 is a potent SIRT2 inhibitor with an IC50 value of 7.0 μM. MIND4-19 can be used for researching Huntington's disease.
  • Potent SIRT2 inhibitor.
  • IC50 value of 7.0 μM for SIRT2.
  • Can be used for researching Huntington's disease.
Catalog No. 50-003-60557 Supplier Medchemexpress LLC Supplier No. HY147233100MG
MIND4-19 is a potent SIRT2 inhibitor with an IC50 value of 7.0 μM. MIND4-19 can be used for researching Huntington's disease.
  • Potent SIRT2 inhibitor.
  • IC50 value of 7.0 μM for SIRT2.
  • Can be used for researching Huntington's disease.